Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AVL9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562278
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562278 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562278 Supplier Invitrogen™ Supplier No. PA562278
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PNHPFQGQYS VSDMKLRFSH SVQNSERGKK IGNVMVTTSR NVVQTGKAVG QSVGGAFSSA KTAMSSWLST FTTSTSQSLT EPPDE Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 95%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen AVL9
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AVL9
Gene Accession No. Q8NBF6
Gene Alias 5830411G16Rik; Avl9; AVL9 cell migration associated; AVL9 homolog (S. cerevisiase); D730049P16Rik; KIAA0241; Late secretory pathway protein AVL9 homolog; mKIAA0241
Gene Symbols AVL9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AVL9. Recombinant protein control fragment (Product #RP-101179).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23080
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.