Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Avobenzone, 97%, Spectrum™ Chemical
SDP

Catalog No. 18600817 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Amber Glass Bottle

A2682, 70356-09-1, C20H22O3

Spectrum™ Chemical Avobenzone is an oil soluble ingredient used in sunscreen products to absorb the full spectrum of UVA rays. Its ability to absorb ultraviolet light over a wider range of wavelengths than many organic sunscreen agents has led to its use in many commercial preparations marketed as ′broad spectrum′ sunscreens. Avobenzone has an absorption maximum of 357 nm. It appears as an odorless, yellowish solid.

Chemical Identifiers

CAS 70356-09-1
Molecular Formula C20H22O3
Molecular Weight (g/mol) 310.39
MDL Number MFCD00210252
InChI Key XNEFYCZVKIDDMS-UHFFFAOYSA-N
IUPAC Name 1-(4-tert-butylphenyl)-3-(4-methoxyphenyl)propane-1,3-dione
SMILES COC1=CC=C(C=C1)C(=O)CC(=O)C1=CC=C(C=C1)C(C)(C)C

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 97%
Grade Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.