Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AVP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA184556
Description
Rabbit anti arginine vasopressin antibody recognizes arginine vasopressin.
This gene encodes a precursor protein consisting of arginine vasopressin and two associated proteins, neurophysin 2 and a glycopeptide, copeptin. Arginine vasopressin is a posterior pituitary hormone which is synthesized in the supraoptic nucleus and paraventricular nucleus of the hypothalamus. Along with its carrier protein, neurophysin 2, it is packaged into neurosecretory vesicles and transported axonally to the nerve endings in the neurohypophysis where it is either stored or secreted into the bloodstream. The precursor is thought to be activated while it is being transported along the axon to the posterior pituitary. Arginine vasopressin acts as a growth factor by enhancing pH regulation through acid-base transport systems. It has a direct antidiuretic action on the kidney, and also causes vasoconstriction of the peripheral vessels. This hormone can contract smooth muscle during parturition and lactation. It is also involved in cognition, tolerance, adaptation and complex sexual and maternal behaviour, as well as in the regulation of water excretion and cardiovascular functions.
Specifications
AVP | |
Polyclonal | |
Unconjugated | |
AVP | |
ADH; antidiuretic hormone; arginine vasopressin; arginine vasopressin (Diabetes insipidus), same as Di (conflicting physical mapping); arginine vasopressin (neurophysin II, antidiuretic hormone, diabetes insipidus, neurohypophyseal); arginine vasopressin-neurophysin II precursor; Arginine-vasopressin; Arg-vasopressin; ARVP; Avp; AVP-neurophysin II precursor; AVP-NPII; AVRP; Copeptin; DI; neurohypophyseal; Neurophysin 2; Neurophysin-I; Neurophysin-II; prepro-arginine-vasopressin-neurophysin II; prepro-AVP-NP II; prepropressophysin; preprovasopressin-neurophysin; Vas; Vasopressin; vasopressin-neurophysin 2-copeptin; vasopressin-neurophysin II-copeptin; vasopressin-neurophysin prepropeptide; VP; Vsp | |
Rabbit | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Immunohistochemistry (Frozen) | |
Conc. Not Determined | |
Whole Serum, PBS with <0.1% sodium azide | |
P01186 | |
AVP | |
Synthetic peptide corresponding to residues CYFQNCPRG of Arginine Vasopression (aa 24-32). | |
RUO | |
24221 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction