Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AVP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112845
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112845 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112845 Supplier Invitrogen™ Supplier No. PA5112845
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AVP Polyclonal Antibody for ELISA, IHC (P), Western Blot

This gene encodes a precursor protein consisting of arginine vasopressin and two associated proteins, neurophysin 2 and a glycopeptide, copeptin. Arginine vasopressin is a posterior pituitary hormone which is synthesized in the supraoptic nucleus and paraventricular nucleus of the hypothalamus. Along with its carrier protein, neurophysin 2, it is packaged into neurosecretory vesicles and transported axonally to the nerve endings in the neurohypophysis where it is either stored or secreted into the bloodstream. The precursor is thought to be activated while it is being transported along the axon to the posterior pituitary. Arginine vasopressin acts as a growth factor by enhancing pH regulation through acid-base transport systems. It has a direct antidiuretic action on the kidney, and also causes vasoconstriction of the peripheral vessels. This hormone can contract smooth muscle during parturition and lactation. It is also involved in cognition, tolerance, adaptation and complex sexual and maternal behaviour, as well as in the regulation of water excretion and cardiovascular functions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AVP
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene AVP
Gene Accession No. P01185, P01186
Gene Alias ADH; antidiuretic hormone; arginine vasopressin; arginine vasopressin (Diabetes insipidus), same as Di (conflicting physical mapping); arginine vasopressin (neurophysin II, antidiuretic hormone, diabetes insipidus, neurohypophyseal); arginine vasopressin-neurophysin II precursor; Arginine-vasopressin; Arg-vasopressin; ARVP; Avp; AVP-neurophysin II precursor; AVP-NPII; AVRP; Copeptin; DI; neurohypophyseal; Neurophysin 2; Neurophysin-I; Neurophysin-II; prepro-arginine-vasopressin-neurophysin II; prepro-AVP-NP II; prepropressophysin; preprovasopressin-neurophysin; Vas; Vasopressin; vasopressin-neurophysin 2-copeptin; vasopressin-neurophysin II-copeptin; vasopressin-neurophysin prepropeptide; VP; Vsp
Gene Symbols AVP
Host Species Rabbit
Immunogen Recombinant Human Vasopressin-neurophysin 2-copeptin protein (126-164AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24221, 551
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.