Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AVPI1 Antibody (OTI1G1), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272121B
Description
AVPI1 Monoclonal specifically detects AVPI1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
AVPI1 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
AVPI1 | |
Full length recombinant protein of human AVPI1 (NP_068378) produced in E.coli. | |
0.1 mL | |
60370.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI1G1 | |
Western Blot, Immunohistochemistry | |
arginine vasopressin-induced 1, arginine vasopressin-induced protein 1, AVP-induced protein 1, PP5395, RP11-548K23.7, vasopressin-induced transcript, VIP32, VIT32 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction