Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AVPI1 Monoclonal Antibody (OTI1G1)
GREENER_CHOICE

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA526567

Catalog No. PIMA526567


Only null left
Add to Cart

Description

Description

AVPI1 Monoclonal Antibody for Western Blot, IHC (P)

May be involved in MAP kinase activation, epithelial sodium channel (ENaC) down-regulation and cell cycling. Target protein is homologous to mouse arginine vasopressin-induced 1 gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

AVPI1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q5T686
AVPI1
Full length human recombinant protein of AVPI1 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI1G1
Unconjugated
AVPI1
arginine vasopressin induced 1; arginine vasopressin-induced 1; arginine vasopressin-induced protein 1; AVPI1; AVP-induced protein 1; NPD013; PP5395; RP11-548K23.7; vasopressin-induced transcript; VIP32; VIT32
Mouse
Affinity Chromatography
RUO
60370
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.