Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AVPI1 Monoclonal Antibody (OTI6C5), TrueMAB™, OriGene
SDP

Catalog No. 501679532
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-9532 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-9532 Supplier OriGene Supplier No. TA802991
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

May be involved in MAP kinase activation, epithelial sodium channel (ENaC) down-regulation and cell cycling.

Specifications

Antigen AVPI1
Applications Western Blot
Classification Monoclonal
Clone OTI6C5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene AVPI1
Gene Accession No. Q5T686
Gene Alias PP5395, VIP32, VIT32
Gene Symbols AVPI1
Host Species Mouse
Immunogen Full length human recombit protein of human AVPI1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60370
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.