Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AVPI1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558041
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558041 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558041 Supplier Invitrogen™ Supplier No. PA558041
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASVVSEPPPW QAPIEARGRK QASANIFQDA ELLQIQGLFQ RSGDQLAEER AQIIWECAGD HRVAEALKR Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 80%.

May be involved in MAP kinase activation, epithelial sodium channel (ENaC) down-regulation and cell cycling. Target protein is homologous to mouse arginine vasopressin-induced 1 gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AVPI1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AVPI1
Gene Accession No. Q5T686
Gene Alias arginine vasopressin induced 1; arginine vasopressin-induced 1; arginine vasopressin-induced protein 1; AVPI1; AVP-induced protein 1; NPD013; PP5395; RP11-548K23.7; vasopressin-induced transcript; VIP32; VIT32
Gene Symbols AVPI1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AVPI1. Recombinant protein control fragment (Product #RP-97065).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60370
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.