Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AVPR1A/AVP Receptor V1a Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981504
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1504 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1504 Supplier Bioss Supplier No. BS11598R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Vasopressin receptor 1a (AVPR1A) is a receptor for arginine vasopressin. The activity of this receptor is mediated by G-proteins which activate a phosphatidyl-inositol-calcium second messenger system. AVPR1A has been involved in social behaviors, including affiliation and attachment.

Specifications

Antigen AVPR1A/AVP Receptor V1a
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene AVPR1A
Gene Accession No. P37288
Gene Alias Antidiuretic hormone receptor 1a; arginine vasopressin receptor 1A; arginine vasopressin receptor V1a; AVPR; AVPR V1a; AVPR1; AVPR1A; SCCL vasopressin subtype 1a receptor; V1a; V1a arginine vasopressin receptor; V1a receptor; V1a vasopressin receptor; V1aR; V1-vascular vasopressin receptor AVPR1A; Vascular/hepatic-type arginine vasopressin receptor; Vasopressin V1a receptor; Vasopressin V1aR
Gene Symbols AVPR1A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human AVP Receptor V1a .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 552
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.