Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AVPR1A Mouse anti-Human, Mouse, Clone: 721CT25.2.1, Abnova™

Catalog No. 89933602 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-602 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-602 Supplier Abnova Corporation Supplier No. MAB12875
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of human AVPR1A.

The protein encoded by this gene acts as receptor for arginine vasopressin. This receptor belongs to the subfamily of G-protein coupled receptors which includes AVPR1B, V2R and OXT receptors. Its activity is mediated by G proteins which stimulate a phosphatidylinositol-calcium second messenger system. The receptor mediates cell contraction and proliferation, platelet aggregation, release of coagulation factor and glycogenolysis. [provided by RefSeq]

Specifications

Antigen AVPR1A
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 721CT25.2.1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of human AVPR1A.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide).
Gene Alias AVPR1
Gene Symbols AVPR1A
Host Species Mouse
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 353-383 of human AVPR1A.
Purification Method Euglobulin precipitation
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 552
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.