Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AVPR2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568479
Description
Predicted to react with Human samples.
Avpr2 is a precursor protein consisting of arginine vasopressin and two associated proteins, neurophysin 2 and a glycopeptide, copeptin. Arginine vasopressin is a posterior pituitary hormone which is synthesized in the supraoptic nucleus and paraventricular nucleus of the hypothalamus. Along with its carrier protein, neurophysin 2, it is packaged into neurosecretory vesicles and transported axonally to the nerve endings in the neurohypophysis where it is either stored or secreted into the bloodstream. The precursor is thought to be activated while it is being transported along the axon to the posterior pituitary. Arginine vasopressin acts as a growth factor by enhancing pH regulation through acid-base transport systems. Avpr2 has a direct antidiuretic action on the kidney, and also causes vasoconstriction of the peripheral vessels. Avpr2 can contract smooth muscle during parturition and lactation. Avpr2 is also involved in cognition, tolerance, adaptation and complex sexual and maternal behaviour, as well as in the regulation of water excretion and cardiovascular functions. The activity of Avpr2 is mediated by G proteins which activate adenylate cyclase.
Specifications
AVPR2 | |
Polyclonal | |
Unconjugated | |
AVPR2 | |
ADHR; antidiuretic hormone receptor; arginine vasopressin receptor 2; Avp2r; AVPR V2; AVPR2; DI1; DIR; DIR3; MGC126533; MGC138386; ND1; NDI; nephrogenic diabetes insipidus; renal-type arginine vasopressin receptor; V2R; V2-renal V2R; Vasopressin receptor V2; vasopressin V2 receptor; VPV2R | |
Rabbit | |
affinity chromatography | |
RUO | |
12000, 554 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O88721, P30518 | |
AVPR2 | |
Synthesized peptide derived from human AVPR2 [aa 1-50]. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction