Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Invitrogen™ AVPR2 Polyclonal Antibody
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109435
Description
Immunogen sequence: IHASLVPGPS ERPGGRRRGR RTGSPGEGAH VSAAVAKTVR MTLVIVVVYV LCWAPFFLVQ LWAAWDPEAP LEGAPFVLLM LLASLNSCTN PWIYASFSSS VSSELRSLLC CARGRTPPSL GPQDESCTTA SSSLAKDTSS.
Avpr2 is a precursor protein consisting of arginine vasopressin and two associated proteins, neurophysin 2 and a glycopeptide, copeptin. Arginine vasopressin is a posterior pituitary hormone which is synthesized in the supraoptic nucleus and paraventricular nucleus of the hypothalamus. Along with its carrier protein, neurophysin 2, it is packaged into neurosecretory vesicles and transported axonally to the nerve endings in the neurohypophysis where it is either stored or secreted into the bloodstream. The precursor is thought to be activated while it is being transported along the axon to the posterior pituitary. Arginine vasopressin acts as a growth factor by enhancing pH regulation through acid-base transport systems. Avpr2 has a direct antidiuretic action on the kidney, and also causes vasoconstriction of the peripheral vessels. Avpr2 can contract smooth muscle during parturition and lactation. Avpr2 is also involved in cognition, tolerance, adaptation and complex sexual and maternal behaviour, as well as in the regulation of water excretion and cardiovascular functions. The activity of Avpr2 is mediated by G proteins which activate adenylate cyclase.
            Specifications
| AVPR2 | |
| Polyclonal | |
| Unconjugated | |
| AVPR2 | |
| ADHR; antidiuretic hormone receptor; arginine vasopressin receptor 2; Avp2r; AVPR V2; AVPR2; DI1; DIR; DIR3; MGC126533; MGC138386; ND1; NDI; nephrogenic diabetes insipidus; renal-type arginine vasopressin receptor; V2R; V2-renal V2R; Vasopressin receptor V2; vasopressin V2 receptor; VPV2R | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 554 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | 
| ELISA, Western Blot | |
| 0.72 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P30518 | |
| AVPR2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 232-371 of human AVPR2 (NP_000045.1). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG | 
Safety and Handling
                                                    
                                                    WARNING: Cancer - www.P65Warnings.ca.gov 
                                                
                                            
                                        
                                    
                                
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction