Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AWAT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564118
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564118 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564118 Supplier Invitrogen™ Supplier No. PA564118
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WVRNWCVWTH IRDYFPITIL KTKDLSPEHN YLMGVHPHGL LTFGAFCNFC T Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.

The protein encoded by this gene belongs to the diacylglycerol acyltransferase family. It esterifies long chain (wax) alcohols with acyl-CoA-derived fatty acids to produce wax esters. Wax esters are enriched in sebum, suggesting that this enzyme plays a central role in lipid metabolism in skin. Consistent with this observation, this protein is predominantly expressed in the sebaceous gland of the skin. [provided by RefSeq, Sep 2009].
TRUSTED_SUSTAINABILITY

Specifications

Antigen AWAT1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AWAT1
Gene Accession No. Q58HT5
Gene Alias Acyl-CoA wax alcohol acyltransferase 1; AWAT1; DGA2; Dgat2l3; diacylglycerol acyltransferase 2; diacyl-glycerol acyltransferase 2; diacylglycerol O-acyltransferase 2-like 3; diacylglycerol O-acyltransferase 2-like protein 3; Long-chain-alcohol O-fatty-acyltransferase 1; wax synthase 1
Gene Symbols AWAT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AWAT1. Recombinant protein control fragment (Product #RP-103862).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158833
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.