Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AWAT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA539003
Description
The protein encoded by this gene belongs to the diacylglycerol acyltransferase family. It esterifies long chain (wax) alcohols with acyl-CoA-derived fatty acids to produce wax esters. Wax esters are enriched in sebum, suggesting that this enzyme plays a central role in lipid metabolism in skin. Consistent with this observation, this protein is predomitly expressed in the sebaceous gland of the skin. [provided by RefSeq, Sep 2009].
Specifications
| AWAT1 | |
| Polyclonal | |
| Unconjugated | |
| AWAT1 | |
| DGA2, DGAT2L3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 158833 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q58HT5 | |
| AWAT1 | |
| A synthetic peptide derived from the internal region of human AWAT1 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction