Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AWAT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564062
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564062 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564062 Supplier Invitrogen™ Supplier No. PA564062
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PLIPAYAFGE TDLYDQHIFT PGGFVNRFQK WFQSMVHIYP CAFYGRGFTK Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 84%.

This gene encodes an enzyme belonging to the diacylglycerol acyltransferase family. This enzyme produces wax esters by the esterification of long chain (or wax) alcohols with acyl-CoA-derived fatty acids. It functions in lipid metabolism in the skin, mostly in undifferentiated peripheral sebocytes. This enzyme may also have acyl-CoA:retinol acyltransferase activities, where it catalyzes the synthesis of diacylglycerols and retinyl esters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AWAT2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AWAT2
Gene Accession No. Q6E213
Gene Alias 11-cis-RE-synthase; 11-cis-specific retinyl-ester synthase; 9430062J17Rik; acyl-CoA retinol O-fatty-acyltransferase; Acyl-CoA wax alcohol acyltransferase 2; ARAT; Awat2; DC4; DGAT2L4; diacylglycerol O-acyltransferase 2-like 4; diacylglycerol O-acyltransferase 2-like protein 4; Diacylglycerol O-acyltransferase candidate 4; hDC4; hWS; Long-chain-alcohol O-fatty-acyltransferase 2; MFAT; Multifunctional O-acyltransferase; mWS; retinol O-fatty-acyltransferase; RGD1565111; Wax synthase; wax synthase 2; WS
Gene Symbols AWAT2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AWAT2. Recombinant protein control fragment (Product #RP-103468).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158835
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.