Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AX 15836, > 98%
GREENER_CHOICE

Catalog No. 584310
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58-431-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 58-431-0 Supplier Tocris Bioscience™ Supplier No. 5843/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective ERK5 inhibitor

Potent and selective ERK5 inhibitor (IC50 = 8 nM). Exhibits >1,000-fold selectivity for ERK5 over a panel of over 200 kinases. Also exhibits selectivity over bromodomain BRD4 (Kd = 3,600 nM). Increases vomocytosis in human macrophages in vitro, as well as in an in vivo zebrafish model of cryptococcal disease, reducing spread of infection.

Chemical Identifiers

CAS 2035509-96-5
Molecular Formula C32H40N8O5S

Specifications

Quantity 10 mg
Solubility Soluble to 100mM in 1eq. HCl and to 100mM in DMSO
Formula Weight 648.78
Purity >98%
Molecular Weight (g/mol) 648.78
Chemical Name or Material 5,11-Dihydro-2-[[2-ethoxy-4-[[4-(4-methyl-1-piperazinyl)-1-piperidinyl]carbonyl]phenyl]amino]-5-meth
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.