Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Axin-1 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176483MFV610
Description
Axin-1 Polyclonal antibody specifically detects Axin-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
Axin-1 | |
Polyclonal | |
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
axin 1, axin-1, AXINaxis inhibitor 1, Axis inhibition protein 1, fused, mouse, homolog of, hAxin, MGC52315 | |
Antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human AXIN1. The immunogen is located within amino acids 620 - 670 of AXIN1. Amino Acid Squence: EKEISRHRRTGHGS | |
0.1 mL | |
Apoptosis, Wnt Signaling Pathway | |
8312 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunocytochemistry | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction