Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axin1 Polyclonal Antibody, Cyanine7, Bioss
SDP

Catalog No. 502280253
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-0253 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-0253 Supplier Bioss Supplier No. BS21732RCY7
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Axin1 Polyclonal antibody specifically detects Axin1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)

Specifications

Antigen Axin1
Applications Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Cyanine7
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene AXIN1
Gene Accession No. O15169, O35625, O70239
Gene Alias AI316800; AXIN; axin 1; Axin1; axin-1; Axis inhibition protein 1; axis inhibitor 1; Fu; fused; fused, mouse, homolog of; GSK-3beta interacting protein rAxin; hAxin; Kb; Ki; kinky; knobbly; LA16c-314G4.3; MGC52315; PPP1R49; Protein Fused; protein phosphatase 1, regulatory subunit 49; rAxin
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Axin1 (301-400).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12005, 79257, 8312
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.