Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axin1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984729
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4729 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4729 Supplier Bioss Supplier No. BS2439R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AXIN1 is a cytoplasmic protein which contains a regulation of G-protein signaling (RGS) domain and a disheveled and axin (DIX) domain and is thought to function as a negative regulator of the WNT signaling pathway that regulates embryonic axis formation. AXIN1 interacts with adenomatosis polyposis coli (APC), beta-catenin, glycogen synthase kinase 3 beta, forming a tetrameric complex resulting in the regulation of the stabilization of beta-catenin. Mutations in the AXIN1 gene have been associated various carcinomas, indicating that it also functions as a tumor suppressor.

Specifications

Antigen Axin1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene AXIN1
Gene Accession No. O15169
Gene Alias AI316800; Axin; axin 1; AXIN1; Axin-1; Axis inhibition protein 1; axis inhibitor 1; Fu; fused; fused, mouse, homolog of; GSK-3beta interacting protein rAxin; hAxin; Kb; Ki; kinky; knobbly; LA16c-314G4.3; MGC52315; PPP1R49; Protein Fused; protein phosphatase 1, regulatory subunit 49; rAxin
Gene Symbols AXIN1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Axin1 .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8312
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.