Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AXIN1 Recombinant Superclonal™ Antibody (11HCLC)

Catalog No. PI710204
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI710204 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI710204 Supplier Invitrogen™ Supplier No. 710204
Add to Cart
Add to Cart

Rabbit Recombinant Superclonal Antibody

Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called 'Recombinant polyclonal antibody', this product is now rebranded as 'Recombinant Superclonal™ antibody'. The physical product and the performance remain unchanged.

AXIN1 is a cytoplasmic protein which contains a regulation of G-protein signaling (RGS) domain and a disheveled and axin (DIX) domain and is thought to function as a negative regulator of the WNT signaling pathway that regulates embryonic axis formation. AXIN1 interacts with adenomatosis polyposis coli (APC), beta-catenin, glycogen synthase kinase 3 beta, forming a tetrameric complex resulting in the regulation of the stabilization of beta-catenin. Mutations in the AXIN1 gene have been associated various carcinomas, indicating that it also functions as a tumor suppressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AXIN1
Applications Western Blot
Classification Recombinant Superclonal
Clone 11HCLC
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene AXIN1
Gene Accession No. O15169
Gene Alias AI316800; AXIN; axin 1; Axin1; axin-1; Axis inhibition protein 1; axis inhibitor 1; Fu; fused; fused, mouse, homolog of; GSK-3beta interacting protein rAxin; hAxin; Kb; Ki; kinky; knobbly; LA16c-314G4.3; MGC52315; PPP1R49; Protein Fused; protein phosphatase 1, regulatory subunit 49; rAxin
Gene Symbols AXIN1
Host Species Rabbit
Immunogen Peptide corresponding to amino acids 319-332 of human Axis Inhibitor Protein 1.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8312
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.