Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Axl Antibody (MM0098-2N33), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212044G
Description
Axl Monoclonal specifically detects Axl in Human, Mouse samples. It is validated for Flow Cytometry, Flow (Cell Surface).Specifications
Axl | |
Monoclonal | |
DyLight 488 | |
AAA61243 | |
AXL | |
Human recombinant Axl EC domain | |
0.1 mL | |
Cell Cycle and Replication, Phospho Specific | |
558 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
MM0098-2N33 | |
Flow Cytometry, Flow (Cell Surface) | |
AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, tyrosine-protein kinase receptor UFO, UFOoncogene AXL | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction