Learn More
Promega AXL Kinase Enzyme System, 1mg
Catalog No. PRV3962 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3962 1 mg
1 options
Catalog No. PRV3962 Supplier Promega Supplier No. V3962
Add to Cart
Add to Cart
Request a Quote
Recombinant human AXL (amino acids 473-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. AXL is a member of the receptor tyrosine kinase family, which has oncogenic potential and is implicated in human myeloid leukemia.
  • Easily Screen and Profile AXL Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C