Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AXL, Mouse anti-Human, Clone: 7E10, Abnova™

Catalog No. 89998902 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-998-902 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-998-902 Supplier Abnova Corporation Supplier No. MAB16174
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifications

Antigen AXL
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 7E10
Conjugate Unconjugated
Dilution ELISA (1:10000) Immunofluorescence (1:200-1:1000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.03% sodium azide).
Gene AXL receptor tyrosine kinase
Gene Alias JTK11/UFO
Gene Symbols AXL
Host Species Mouse
Immunogen Purified recombinant hIgGFc tag fusion fragment corresponding to extracellular domain of human AXL from HEK293 cell line.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 558
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.