Learn More
AXL Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | AXL |
|---|---|
| Concentration | 0.21 mg/mL |
| Applications | Flow Cytometry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-8562
![]()
|
Proteintech Group Inc
131961AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody detects a single band at 140 kDa, which is considered to represent the auto-phosphorylated form of endogenous Axl.
AXL, a member of the AXL/UFO subfamily of Tyr protein kinases, may function as a signal transducer between specific cell types of mesodermal origin. This Type I membrane protein has transforming potential in patients with chronic myeloproliferative disorder or chronic myelocytic leukemia. The protein contains 2 putative fibronectin type III domains and 2 putative immunoglobulin-like C2-type domains.Specifications
| AXL | |
| Flow Cytometry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P30530, Q00993 | |
| 26362, 558 | |
| AXL Fusion Protein Ag3898 | |
| Primary | |
| -20°C | |
| AXL |
| 0.21 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| AXL, AXL oncogene, AXL receptor tyrosine kinase, JTK11, UFO | |
| AXL | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.