Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Axotrophin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537969
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537969 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537969 Supplier Invitrogen™ Supplier No. PA537969
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with human and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

The function of protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Axotrophin
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene MARCH7
Gene Accession No. Q9WV66
Gene Alias AXO; AXOT; Axotrophin; C3HC4 7; DKFZP586F1122; E3 ubiquitin-protein ligase MARCH7; gene trap ROSA b-geo 17; Gtrgeo17; MARCH7; MARCHF7; MARCH-VII; membrane associated ring finger 7; membrane associated ring-CH-type finger 7; membrane-associated ring finger (C3HC4) 7; membrane-associated ring finger (C3HC4) 7, E3 ubiquitin protein ligase; membrane-associated RING finger protein 7; Membrane-associated RING-CH protein VII; RING finger protein 177; RING-type E3 ubiquitin transferase MARCH7; RNF177
Gene Symbols MARCH7
Host Species Goat
Immunogen Peptide with sequence C-RTLQAHMEDLETSED, from the internal region of MARCH7 (aa 677-692).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57438
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.