Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Axotrophin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115545
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115545 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115545 Supplier Invitrogen™ Supplier No. PA5115545
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MARCH7.

The function of protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Axotrophin
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene MARCH7
Gene Accession No. Q9H992
Gene Alias AXO; AXOT; Axotrophin; C3HC4 7; DKFZP586F1122; E3 ubiquitin-protein ligase MARCH7; gene trap ROSA b-geo 17; Gtrgeo17; MARCH7; MARCHF7; MARCH-VII; membrane associated ring finger 7; membrane associated ring-CH-type finger 7; membrane-associated ring finger (C3HC4) 7; membrane-associated ring finger (C3HC4) 7, E3 ubiquitin protein ligase; membrane-associated RING finger protein 7; Membrane-associated RING-CH protein VII; RING finger protein 177; RING-type E3 ubiquitin transferase MARCH7; RNF177
Gene Symbols MARCH7
Host Species Rabbit
Immunogen A synthesized peptide derived from human MARCH7(Accession Q9H992), corresponding to amino acid residues Y67-S117.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64844
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.