Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AXUD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100400
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100400 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100400 Supplier Invitrogen™ Supplier No. PA5100400
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total AXUD1.

Axin, an important regulator of beta-catenin, is frequently mutated in human hepatocellular carcinomas (HCCs), and transduction of the wild-type Axin gene (AXIN1) induces apoptosis in HCC cells as well as in colon cancer cells. A novel gene, AXUD1 (AXIN1 up-regulated), has enhanced expression in response to exogenously expressed AXIN1. It is expressed in all human tissues examined, most abundantly in lung, placenta, skeletal muscle, pancreas and leukocytes. Data suggests that AXUD1 may have a tumor-suppressor function in those organs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AXUD1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CSRNP1
Gene Accession No. P59054, Q96S65
Gene Alias 4931429D10Rik; AXIN1 up-regulated 1; Axin-1 up-regulated gene 1 protein; AXUD1; Csrnp1; CSRNP-1; cysteine and serine rich nuclear protein 1; cysteine/serine-rich nuclear protein 1; cysteine-serine-rich nuclear protein 1; DKFZp566F164; FAM130B; Protein URAX1; Taip3; TAIP-3; TGF-beta induced apoptosis protein 3; TGF-beta-induced apoptosis protein 3; URAX1; URAX1 protein
Gene Symbols CSRNP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CSRNP1(Accession Q96S65), corresponding to amino acid residues S486-H536.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 215418, 363165, 64651
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.