Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AXUD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566174
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566174 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566174 Supplier Invitrogen™ Supplier No. PA566174
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FAQEQARARHE KLRQRLKEEK LEMLQWKLSA AGVPQAEAGL PPVVDAIDDA SVEEDLAVAV AGGRLEEVS Highest antigen sequence identity to the following orthologs - mouse 76%, rat 79%.

Axin, an important regulator of beta-catenin, is frequently mutated in human hepatocellular carcinomas (HCCs), and transduction of the wild-type Axin gene (AXIN1) induces apoptosis in HCC cells as well as in colon cancer cells. A novel gene, AXUD1 (AXIN1 up-regulated), has enhanced expression in response to exogenously expressed AXIN1. It is expressed in all human tissues examined, most abundantly in lung, placenta, skeletal muscle, pancreas and leukocytes. Data suggests that AXUD1 may have a tumor-suppressor function in those organs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AXUD1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CSRNP1
Gene Accession No. Q96S65
Gene Alias 4931429D10Rik; AXIN1 up-regulated 1; Axin-1 up-regulated gene 1 protein; AXUD1; Csrnp1; CSRNP-1; cysteine and serine rich nuclear protein 1; cysteine/serine-rich nuclear protein 1; cysteine-serine-rich nuclear protein 1; DKFZp566F164; FAM130B; Protein URAX1; Taip3; TAIP-3; TGF-beta induced apoptosis protein 3; TGF-beta-induced apoptosis protein 3; URAX1; URAX1 protein
Gene Symbols CSRNP1
Host Species Rabbit
Immunogen Recombinant Human AXUD1. Recombinant protein control fragment (Product #RP-106858).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64651
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.