Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ 1000 μL Universal Pipetter Tips: Bevelled
GREENER_CHOICE

Catalog No. 14222690 Shop All Axygen Products
Click to view available options
Sterility:
Non-sterile
Sterile
Color:
Blue
Clear
Format:
Bulk
Rack

Compatible with most popular brands of pipettes

Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.

  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Beveled
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 100 to 1000 μL
Sterility Non-sterile
Color Blue
Autoclavable Autoclavable
Clean Claims DNase, RNase, and Pyrogen free
Format Bulk
Tip Style Beveled, Universal
Packaging 1,000 Tips/Pk., 5 Packs/Cs.
For Use With (Equipment) Universal Fit
Material Polypropylene
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.