Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ 1250μL Universal Pipette Tips

Catalog No. 14222216
Click to view available options
Packaging:
Bulk
Rack

Matrix compatible tips in clear color

  • Matrix-style tips fits Matrix Impact™ and Impact2™ brand pipettors
  • Maxymum Recovery™ tips allows complete retrieval of precious samples
  • Optimized tip bevel reduces sample hang-up and improves pipetting accuracy
  • Presterilized tips are exposed to e-beam radiation for complete security without degrading the plastic or performance
  • Volume range: 15 to 1250μL
  • Available in bulk or racked formats

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1250 μL
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Certifications/Compliance RNase, DNase-free
Packaging Rack
For Use With (Equipment) Matrix™ Impact™ Pipette
Material Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.