Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Axygen™ 200μL Universal Pipette Tips: 200μL, Beveled
Designed to fit a wide variety of single and multi-channel pipettes.
Supplier: Axygen™ T200C
Description
- Optimized tip bevel reduces sample hang-up, improves pipetting accuracy
- Wide bore tips of orifice size: 0.062 in. (1.52 mm) makes it ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
- Graduation marks allow a visual confirmation of pipettor draw volume
- Presterilized tips are e-beam irradiated to prevent product degradation
- Clean-room manufacturing environment certifies products safe from RNase, DNase and pyrogen safe
- Also available with Maxymum recovery, which allows complete retrieval of precious samples, saves costly reagents
Order Info
Available in bulk, racked or stack rack formats.
Specifications
1 to 200 μL | |
Clear | |
DNase, RNase, DNA, and PCR inhibitor Free | |
Universal, Beveled | |
Polypropylene |
Non-sterile | |
Autoclavable | |
Bulk | |
Universal Fit |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction