Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ 384-well Skirted PCR Microplates

Catalog No. p-4102748 Shop All Axygen Products
Click to view available options
Color:
Clear
White
Volume (Metric) Well:
50 μL
55 μL
Includes:
Single notch

Single and double notch 384-well PCR plates

Sealing Options: Axymats, heat sealing, adhesive films and foils.

  • Available in 50μL and 55μL
  • Single and double notch corner

Skirted (50μL)

  • Application/Compatibility: standard thermal cycling
  • Robotic-friendly full skirt design
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.
Skirted (55μL)
  • Application/Compatibility: standard thermal cycling, fully ABI compatible for qPCR and sequencing
  • Robotic-friendly full skirt design
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.
Skirted, RigidPlate (50μL)
  • Robotic-friendly rigid plate with full skirt
  • Application/Compatibility: standard thermal cycling, ABI sequencing, automation applications
  • Packaging Description: 10 plates/Pk., 2 packs/sleeve, 5 sleeves/Cs.
Skirted, elevated wells
  • Application/Compatibility: standard thermal cycling, ABI sequencing, qPCR
  • Raised wells allow 55μL volume
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.

Colors available by special order.

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance RNase, DNase-free
Format Standard
Lid Without Lid
Array 16 x 24
Color White
For Use With (Equipment) For Roche light cycler 480
Material Polypropylene
Volume (Metric) Well 55 μL
No. of Wells 384
Volume (Metric) Working 30 μL
Sterility Non-sterile
Well Shape Round
For Use With (Application) PCR
Includes Single notch
No. per Case 50/Cs.
No. per Pack 10/Pk.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.