Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ 96-Well Half-Skirt PCR Microplates

Catalog No. 14223337
Click to view available options
Color:
Orange
Yellow
This item is not returnable. View return policy
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
This item is not returnable. View return policy
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit

Half-skirt amplification plate with single notch

PCR™ products specifically designed for use within PCR thermal transfer and real time PCR applications. A variety of colors are offered to help organize multi-step PCR processes.

  • 96-well PCR half-skirt amplification plate with a single notch
  • Working volume is 300μL
  • Available in a variety of colors
  • Sold 10 plates/unit; 5 units/Cs.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance RNase, DNase-free
Format Standard
Array 8 x 12
Color Orange
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Well Shape Round
For Use With (Application) PCR thermal transfer, RT-PCR applications
Quantity 50/Cs.
Well Color Clear
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.