Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Axygen™ 96-Well Low Profile PCR Microplates

Click to view available options
Color:
Clear
White
Skirt Style:
Half
Non-skirt
Description
Constructed with ultra-thin and consistent wall thickness to allow precise thermal transfer for optimum results. Corning™ Axygen™ 96-well PCR Microplates are the ideal option for use in PCR thermal transfer and Real-Time PCR applications. The
Axygen™ 96 well low profile PCR microplate is designed to fit ABI Real Time PCR thermal cyclers and sequencers. Low profile design results in faster and more efficient PCR reactions. Ultra-thin and consistent wall thickness allows precise thermal transfer for optimum results. Can be sealed with 8/12 strip caps, Axymat or sealing film.
RNase-/DNase-free Nonpyrogenic Nonsterile Manufactured with 99.9% pure virgin polypropylene 96-Well Semi-Skirted Low Profile PCR Plate Optimized for Fast PCR Designed for ABI Thermal Cyclers Application/Compatibility: ABI Fast PCR 7500, 7900 9800, Veriti and StepOne

Specifications
Specifications
Barcode | No Barcode |
Certifications/Compliance | RNase-/DNase-free |
Format | Standard |
Lid | Without Lid |
Array | 8 x 12 |
Packaging | 100 Cases, 25 Pk. |
Product Line | Axygen |
Color | Clear |
Product Type | Standard top |
For Use With (Equipment) | ABI thermal cyclers, ABI Fast PCR 7500, 7900 9800, Veriti and StepOne |
Show More |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction