Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ 96-Well 100μL PCR Microplates
GREENER_CHOICE

Catalog No. p-4104931
Click to view available options
Barcode:
Includes Barcode
No Barcode
This item is not returnable. View return policy
This item is not returnable. View return policy

Constructed with ultra-thin and consistent wall thickness to allow precise thermal transfer for optimum results. Corning™ Axygen™ 96-well 100μL PCR Microplates are specifically designed to be compatible with Roche Light Cycler 480.

Axygen™ 96 well PCR microplate is designed to fit Roche Light Cycler 480. Ultra-thin and consistent wall thickness allows precise thermal transfer for optimum results. Includes Ultra Clear sealing film and printed alphanumeric grid for ease of well identification.


  • RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Manufactured with 99.9% pure virgin polypropylene
  • Includes Sealing Films
  • TRUSTED_SUSTAINABILITY

    Specifications

    Barcode Includes Barcode
    Certifications/Compliance RNase-, DNase-free
    Format Standard
    Lid Without Lid
    Array 8 x 12
    Color White
    For Use With (Equipment) Roche Light Cycler 480
    Material Polypropylene
    Volume (Metric) Well 100 μL
    No. of Wells 96
    Sterility Non-sterile
    Well Shape Round
    For Use With (Application) PCR thermal transfer, RT-PCR applications
    Includes Ultra Clear sealing film, bar codes, printed alphanumeric grid
    No. per Case 50/Cs.
    No. per Pack 10/Pk.
    Show More Show Less
    WARNING: Cancer - www.P65Warnings.ca.gov
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.