Learn More
Axygen™ AxyMats™ Sealing Mats for 384 Well Deep Well Plates
Catalog No. p-4879159 Shop All Axygen Products
Click to view available options
Well Shape:
Round
Square
Minimize evaporation in wide range of temperatures in PCR and storage applications
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications. AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved. They can be reused following a bleach wash and ethanol rinse protocol.
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
Material Silicone
Sterility Non-sterile
Autoclavable Autoclavable
No. of Wells 384
No. per Case 50/Cs.
No. per Pack 10/Pk.
Product Type Sealing Mat
Well Shape Square
Plate Compatibility 384-well Deep Well Plate with Square Wells
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.