Learn More
Axygen™ Axyprep™ MAG Plant Genomic DNA Extraction Kit
Shop All Axygen ProductsDescription
The AxyPrep MAG Plant gDNA Extraction Kit is a magnetic beads-based purification system that allows rapid and reliable isolation of high quality gDNA from a wide variety of plant samples. Up to 96 samples (50 mg fresh tissue per sample or 15 mg of dried tissue per sample) can be processed in less than one hour. The AxyPrep MAG Plant gDNA Extraction Kit efficiently eliminates polysaccharides, phenolic compounds and enzyme inhibitors with no organic extractions. This kit is adaptable for use on major liquid handling workstations for high throughput processing.
- Simplified extraction process
- Non-alcohol-based purification
- Compatible with genomics downstream applications
- High quality, high-yield DNA
- Manual and automation friendly
Specifications
Specifications
| For Use With (Application) | DNA extraction and purification |
| Includes | MAG-A1 Particles 250μL; PCL Buffer 8 mL; PDB Buffer 7 mL; HSW Buffer 4 mL; MB Elution Buffer 1.5 mL; Protocol Manual 1 |
| No. of Reactions | 10 Preps |
| Product Type | MAG Plant Genomic DNA Extraction Kit |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.