Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Axygen™ AxyPrep™ Mag Plasmid Kits
Shop All Axygen Products
Click to view available options
No. of Reactions:
384 Preps
96 Preps
Description
The AxyPrep MAG Plasmid purification kit utilizes a magnetic beads-based technology for high throughput purification of plasmid DNA from Escherichia coli cells. AxyPrep MAG Plasmid Kit can also be used with fosmid and BAC vector-based constructs ranging from 5 Kb to 150 Kb. The system consists of steps of pelleting cells, resuspension, lysis, neutralization, DNA binding, ethanol washing, and plasmid elution. The protocol is automation-compatible.
Specifications
Specifications
| Format | 96-well Plate, Tube |
| For Use With (Application) | DNA sequencing, PCR amplification, restriction Enzyme digestion |
| No. of Reactions | 384 Preps |
| Product Type | Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction