Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ AxyPrep™ Mag Plasmid Kits

Catalog No. p-4265677 Shop All Axygen Products
Click to view available options
No. of Reactions:
384 Preps
96 Preps

Catalog No. 14-223-167

Supplier: Axygen™ MAGPM

Only null left
Add to Cart
Add to Cart

Utilizes magnetic bead technology for high-throughput purification of plasmid DNA from E.coli cells

The AxyPrep MAG Plasmid purification kit utilizes a magnetic beads-based technology for high throughput purification of plasmid DNA from Escherichia coli cells. AxyPrep MAG Plasmid Kit can also be used with fosmid and BAC vector-based constructs ranging from 5 Kb to 150 Kb. The system consists of steps of pelleting cells, resuspension, lysis, neutralization, DNA binding, ethanol washing, and plasmid elution. The protocol is automation-compatible.

TRUSTED_SUSTAINABILITY

Specifications

Format 96-well Plate, Tube
For Use With (Application) DNA sequencing, PCR amplification, restriction Enzyme digestion
No. of Reactions 384 Preps
Product Type Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.