Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Axygen™ Biomek™ FX/NX Robotic Tips
Click to view available options
Volume (Metric):
20 μL
30 μL
50 μL
250 μL
Sterility:
Non-sterile
Sterile
Tip Style:
Automation, Fine Point
Automation, Round
Automation, Wide Orifice
Description
The Axygen brand range of products includes a comprehensive line of tips for Beckman Coulter automated liquid handling systems. These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. The pipette tips are also available with MAXYMum Recovery, a unique low-retention technology that improves tip performance and maximizes the use of your precious samples.
- Available in racked, racked sterile and with aerosol barrier (filtered)
- Compatible with Beckman Biomek FX and Beckman Multimek instruments with an AP96 head
- Also compatible with Span-8 pod when Liquid Level Sensing function is turned off
- Unsurpassed straightness to ensure tip enters the well smoothly, every time
- 384/1536-well compatible to meet your most demanding applications
- Wide bore tips are ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
- Fine Point improves accuracy and precision at lower sample volumes
- Maxymum recovery™ tips reduce sample loss, improves reproducibility and accuracy, minimizes usage of precious samples and reagents

Specifications
Specifications
Volume (Metric) | 250 μL |
Filtered | Non-filtered |
Sterility | Non-sterile |
Color | Clear |
Autoclavable | Not Autoclavable |
Certifications/Compliance | ANSI |
Clean Claims | DNase, RNase and Pyrogen free |
For Use With (Application) | Non-pyrogenic, used for stringent specifications under strict process controls |
Format | Rack, 96 |
Tip Style | Automation, Round |
Show More |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction