Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Disposable V-Bottom Reagent Reservoirs
GREENER_CHOICE

Catalog No. p-4909860
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
Sterility:
Non-sterile
Sterile
Packaging:
100/Cs.
200/Cs.
Divided; 100/Cs.
Divided; 200/Cs.
Divided; Individually Wrapped; 100/Cs.
Individually Wrapped; 100/Cs.

Designed to accommodate all multichannel pipettor

Axygen disposable reservoirs are designed to accommodate all multichannel pipettors. Four corners and a sharp V-bottom shape ensure maximum sample retrieval. All reservoirs are white.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI, RNase/DNase-free, non-pyrogenic
Color White
Capacity (Metric) 25 mL
Material Polystyrene
Sterility Non-sterile
Type Reagent Reservoir
Packaging Divided; 100/Cs.
Disposable Single-use
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.