Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Axygen™ Disposable V-Bottom Reagent Reservoirs
Shop All Axygen Products

Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
Sterility:
Non-sterile
Sterile
Packaging:
100/Cs.
200/Cs.
Divided; 100/Cs.
Divided; 200/Cs.
Divided; Individually Wrapped; 100/Cs.
Individually Wrapped; 100/Cs.
Description
Designed to accommodate all multichannel pipettor
Axygen disposable reservoirs are designed to accommodate all multichannel pipettors. Four corners and a sharp V-bottom shape ensure maximum sample retrieval. All reservoirs are white.

Specifications
Specifications
Certifications/Compliance | ANSI, RNase/DNase-free, non-pyrogenic |
Color | White |
Capacity (Metric) | 25 mL |
Material | Polystyrene |
Sterility | Sterile |
Type | Reagent Reservoir |
Packaging | Divided; 200/Cs. |
Disposable | Single-use |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction