Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ DNA Marker Kit

Catalog No. 14223050 Shop All Axygen Products
Click to view available options
No. of Bands:
12
13
This item is not returnable. View return policy

Catalog No. 14-223-050

Supplier: Axygen™ MDNA100BP

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

A range of DNA markers to serve as standards for wide range of applications

Room temperature stability and long shelf life enables benchtop storage for easy access, less refrigerator usage and reduced shipping costs. All five markers create precise sharp bands with locations that are consistent under conventional electrophoresis conditions. Sharp bands allow precise detection by gel imaging systems and software for accurate molecular weight calculations. The markers are robust and can be used for fast electrophoresis runs (10 to 15 min).

  • Allow sizing of a wide range of DNA fragments from 100 to 10,000bp
  • Stable at ambient temperature
  • Long shelf life of two years at ambient temperature
  • Crisp and precise band patterns
  • Ready-to-use
  • Formulated with Bromophenol Blue for easy loading

100bp and 1kb Ladders

  • Each vial has 500μL of the marker sufficient for up to 100 lanes
  • The most popular markers used for DNA fragment size determination
  • Allow sizing from 100 to 10,000bp
  • Supplied at a concentration of 50μg/500μL (1μg/10μL)
Low, high and broad range markers
Come in vials of 600μL and are sufficient for up to 125 lanes
Offer an alternative to expensive brands
Offer the widest sizing range, from 125 to 10,000bp
Supplied at a concentration of 1μg/10μL

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 μg/10 μL
Product Type DNA Molecular Weight Marker/Ladder
Formulation 50μg/500μL Ladder DNA marker, Bromophenol Blue
For Use With (Application) Gel electrophoresis
Kit Contents Two ready-to-use DNA size marker
No. of Bands 12
Quantity 500 μL
Storage Requirements Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.