Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ FDSS™ Robotic Tips
GREENER_CHOICE

Catalog No. p-4102775 Shop All Corning Products
Click to view available options
For Use With (Equipment):
Hamamatsu FDSS 7000 System

For Hamamatsu FDSS series systems

  • For 384-Well plates
  • Tip color: Black
  • Capacity: 50μL
  • Packaging: 384 Tips/rack; 10 Racks/unit; 5 Units/Cs.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 50 μL
Filtered Non-filtered
Sterility Non-sterile
Color Black
Autoclavable Autoclavable
Certifications/Compliance Non-ANSI
Format Rack
Tip Style Automation, Round
Packaging SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) Hamamatsu FDSS 7000 System
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.