Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Gel Cutting Tips: 1.1 x 6.5mm

Catalog No. p-8015619 Shop All Axygen Products
Click to view available options
Format:
Bulk
Rack

Universal fit for research grade pipettes.

Axygen™ Gel Cutting Tips are designed to remove bands from agarose gel. These polypropylene tips are nonpyrogenic and RNase-/DNase-free.

 

  • Designed for removing bands from agarose gel
  • One handed operation
  • Push button gel release
  • Ejector button tip release
  • Avoid cross contamination
  • Tips fit standard 1000 μL pipettors
  • These polypropylene tips are nonpyrogenic and RNase-/DNase-free.
  • Nonpyrogenic
  • Nonsterile
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1000 μL
Sterility Non-sterile
Color Clear
Length (Metric) 1.1 x 6.5 mm
Clean Claims DNase, RNase Free
Format Bulk
Tip Style Gel Loading
Packaging 250 Tips/Pk, 10 Packs/Cs
Graduated Yes
Material Polypropylene
Graduations 2, 5μL
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.