Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ LightCycler 480 Compatible PCR Plates

Catalog No. p-4371198 Shop All Axygen Products
Change view
Click to view available options
Color:
White
Includes:
Barcode
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Includes
14-223-390 White Barcode
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-223-390 Supplier Axygen™ Supplier No. PCR384LC480WNFBC
Only null left
Add to Cart
Edge
Add to Cart

Designed to fit the Roche LightCycler™ 480

The new 480 plate offers another great solution for real-time PCR. Thin walls coupled with the 480 platform deliver real-time PCR results in less than 40 minutes. Every well of every plate undergoes an automated QC inspection
for microscopic holes, which gives you peace of mind when handling your valuable samples.

  • Optimized for the new plate-based Roche Light Cycler 480
  • Ideal for real-time PCR, white polypropylene magnifies signal intensity and prevents cross-talk between wells
  • Thin walls reduce cycle times
  • SBS standard dimensions
  • Black printing of alphanumerics for easily identifying wells and orienting plates
  • 100% automated QC for microscopic holes
  • Working volume: 3μL

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance RNase, DNase-free
Format Standard
Array 16 x 24
Color White
For Use With (Equipment) For Roche light cycler 480
No. of Wells 384
Volume (Metric) Working 3 μL
For Use With (Application) RT-PCR
Includes Barcode
Quantity 50/Cs.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.