Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ PCR Tubes with 0.5 mL Flat Cap

Catalog No. 14222294 Shop All Axygen Products
Change view
Click to view available options
Color:
Assorted
Clear
Product Type:
Maxymum Recovery PCR Tube
Thin Wall PCR Tube
Quantity:
1000/Pk., 10Pk./Cs.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type Quantity
14-222-294 Clear Maxymum Recovery PCR Tube 1000/Pk., 10Pk./Cs.
14-222-290 Assorted Thin Wall PCR Tube 1000/Pk., 10Pk./Cs.
14-222-292 Clear Thin Wall PCR Tube 1000/Pk., 10Pk./Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 14-222-294 Supplier Axygen™ Supplier No. PCR05LC
This item is not returnable. View return policy

Thin and consistent wall thickness for precise thermal transfer

Quantitative PCR compatible tubes for a wide range of Real-Time PCR instruments.

  • Tubes with caps sealed full-length; insertion cap ensures tight fit, eliminates sample evaporation
  • Choose from a wide variety of colors
  • Certified RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Manufactured from polypropylene

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance RNase, DNase, pyrogen-free
Color Clear
Volume (Metric) 0.5 mL
Quantity 1000/Pk., 10Pk./Cs.
Closure Material Polypropylene
For Use With (Equipment) Standard 96-well blocks of real-time PCR instruments and thermal transfer applications
Shape Conical
Sterility Non-sterile
Material Polypropylene
Closure Type Flat Cap
Format Tube
Product Type Maxymum Recovery PCR Tube
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.