Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Axygen™ Single Well High Profile Reagent Reservoirs
Click to view available options
Capacity (Metric):
240 mL
282 mL
290 mL
Sterility:
Non-sterile
Sterile
Channels:
8
12
96
384
Description
Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.
- Durable and cost effective
- Various formats for multiple applications

Specifications
Specifications
Certifications/Compliance | ANSI/SLAS, RNase/DNase-free, non-pyrogenic |
Color | Clear |
Capacity (Metric) | 240 mL |
Material | Polypropylene |
Sterility | Non-sterile |
Type | Reagent Reservoir |
No. per Case | 25/Cs. |
No. per Pack | 25/Pk. |
Packaging | 25/Cs. |
Channels | 96 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction