Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Tip Refill System, Empty Racks

Catalog No. p-4909374 Shop All Axygen Products
Click to view available options
Model:
RFL-R
RFL-R-1200
Packaging:
10 Racks/Pack; 3 Packs/Case
10 Racks/Pack; 5 Packs/Case

Catalog No. 14-222-439

Supplier: Axygen™ RFLR

Only null left
Add to Cart
Add to Cart

Easy-to-load refill racks for Axygen Tip Refill System. For use with P2/P10 Gilson-Style and 200μL Universal fit inserts.

Axygen™ Tip Refill System contains durable refill rack that can be easily refilled by hand and is autoclavable. These translucent Racks includes color-coded inserts for tip identification and are certified RNase-/DNase free. They are available in different colors and are packaged in bulk or racks.

  • RFL-R is suitable for both the 10 μL and 200 μL refill inserts
  • RFL-R-1200 is suitable for 1200 μL refill inserts
  • Certified to be RNase-/DNase-free
  • Nonpyrogenic
  • Translucent rack allows tips to be easily seen
  • Color-coded insert allows for easy identification of tip
TRUSTED_SUSTAINABILITY

Specifications

Product Type Empty Refill Rack with Lid
Color Clear
Sterility Non-sterile
For Use With (Equipment) P2/P10 Gilson-Style and 200μL Universal fit inserts
Model RFL-R
Packaging 10 Racks/Pack; 5 Packs/Case
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.