Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Automation Tips for Zymark™/Caliper, 25 μL
GREENER_CHOICE

Catalog No. 14222915 Shop All Axygen Products
Change view
Click to view available options
Sterility:
Non-sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
14-222-915 Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-222-915 Supplier Axygen™ Supplier No. ZT384CR
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compatible with automated liquid handling systems. Axygen™ Automation Tips for Zymark™/Caliper, 25μL are non-filtered pipette tips in 384-well racks. Constructed of clear polypropyle under strict controls to ensure quality. Options include sterile and non-sterile tips.

  • Tip format: 96 tip and 384 tip
  • Tip volume range: 25 μL to 200 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile
    - Standard or Axygen™ Maxymum Recovery™ surface
  • Compatible heads: HV96, CAS-4200-1 (20 - 150 μL/sec.), LV96, LV384
  • Rack format: Non-ANSI (96) and ANSI/SLAS (384)
  • Rack material: Polypropylene
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 30 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Rack Color Light Green
Autoclavable Autoclavable
Clean Claims DNase, RNase Free
For Use With (Application) Non-pyrogenic
Format Rack
Tip Style Automation, Round
Packaging Non-SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) Zymark/Caliper RapidPlate 96/384, SciClone, Zephyr LV96, LV384 Head
Material Polypropylene
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.